Learn More
Tocris Bioscience™ [Nle4,D-Phe7]-α-MSH
GREENER_CHOICE
Catalog No. 30131
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
30-131 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 30-131 Supplier Tocris Bioscience™ Supplier No. 3013/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Melanocortin receptor agonist
[Nle4,D-Phe7]-α-MSH is a synthetic analog of α-MSH that is an agonist at melanocortin receptors (Ki values are 0.085, 0.4, 3.8 and 5.1 nM for MC1, MC3, MC4 and MC5 receptors respectively).
Specifications
Quantity 1 mg