Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Nogo-66 (1-40)

Catalog No. 19841
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-841 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-841 Supplier Tocris Bioscience™ Supplier No. 1984/1
This item is not returnable. View return policy

Competitive antagonist for Nogo-66 receptor; promotes neuron regeneration

Nogo-66 (1-40) is a peptide fragment corresponding to residues 1 - 40 of Nogo-66, the domain of the myelin protein Nogo that inhibits axonal outgrowth. Acts as a competitive antagonist at the Nogo-66 receptor (NgR); blocks Nogo-66- and CNS myelin-induced inhibition of axonal growth, but does not reduce myelin-associated glycoprotein (MAG) inhibition of neurite outgrowth in vitro. Promotes regeneration of hemisected spinal axons and locomotor recovery following spinal injury in vivo.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.