Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
Obtustatin is a highly potent integrin α1β1 inhibitor (IC50 = 0.8 nM for α1β1 binding to type IV collagen). Selective for α1β1 over α2β1, αIIbβ3, αvβ3, α4β1, α5β6, α9β1 and α4β7. Inhibits FGF2-stimulated angiogenesis in the chicken chorioallantoic model. Displays antitumor efficacy in a synergistic mouse model of Lewis lung carcinoma; blocks human melanoma growth in nude mice.
Specifications
Specifications
| Purity | 95% |
| CAS | 404882-00-4 |
| Quantity | 100 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?

