Learn More
Tocris Bioscience™ Oxytocin
GREENER_CHOICE
Catalog No. 19101
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
19-101 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-101 Supplier Tocris Bioscience™ Supplier No. 1910/1
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Involved in uterine contraction and lactation
Oxytocin is a neurohypophyseal peptide. Stimulates uterine contraction and lactation.
Specifications
Quantity 1 mg