Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PACAP 1-27
GREENER_CHOICE

Catalog No. 1183100U
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
1183100U 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 1183100U Supplier Tocris Bioscience™ Supplier No. 1183/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent stimulator of adenylyl cyclase

PACAP 1-27 is an endogenous neuropeptide showing considerable homology with vasoactive intestinal peptide (VIP). Potently stimulates adenylyl cyclase.

Specifications

Purity 0.95
Content And Storage Store at -20°C
Quantity 100 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.