Learn More
Tocris Bioscience™ Pam3CSK4
GSA_VAGREENER_CHOICE
Catalog No. 46331
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
46-331 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 46-331 Supplier Tocris Bioscience™ Supplier No. 4633/1
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
TLR1/2 agonist; induces cytokine production
Pam3CSK4 is a toll-like receptor 1/2 (TLR1/2) agonist; induces production of TNF-α and IL-6 in macrophages. Stimulates phosphorylation of p100/p110 and p60 in granulocytic-differentiated HL-60 cells. Promotes differentiation of naive CD4+ T cells into Th17 cells.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 1 mg