Learn More
Tocris Bioscience™ Pepstatin A
GREENER_CHOICESDP
Catalog No. 119010 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
Catalog No. Quantity
119010 10 mg
1 options
This item is not returnable. View return policy
Catalog No. 119010 Supplier Tocris Bioscience™ Supplier No. 1190/10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective and high affinity cathepsin D inhibitor
Pepstatin A is an irreversible inhibitor of aspartic proteases. Inhibits lysosomal proteases and interferes with autolysosomal digestion when used in combination with E 64d (Cat. No. 4545).
Specifications
Molecular Weight (g/mol) Observed M.W.: 685.91; Theoretical M.W.: 0
Quantity 10 mg