Learn More
Tocris Bioscience™ Phalloidin
GREENER_CHOICE
Catalog No. 45351
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
45-351 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 45-351 Supplier Tocris Bioscience™ Supplier No. 4535/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Promotes actin polymerization
Phalloidin is a promotes actin polymerization. Decreases dissociation rate constant for actin subunits from filament ends; lowers critical concentration for polymerization.
Specifications
Quantity 1 mg