Learn More
Tocris Bioscience™ Phrixotoxin 3
GREENER_CHOICE
Catalog No. 4914100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
49-141-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 49-141-00U Supplier Tocris Bioscience™ Supplier No. 4914/100U
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent blocker of NaV1.2, NaV1.3 and NaV1.5 channels
Phrixotoxin 3 is a potent blocker of voltage-gated sodium channels (IC50 values are 0.6, 42, and 72 nM for NaV1.2, NaV1.3 and NaV1.5 respectively). Blocks inward sodium currents in a voltage-dependent manner.
Specifications
Purity 95%