Learn More
Tocris Bioscience™ PLP (139-151)
GREENER_CHOICE
Catalog No. 25671
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
25-671 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 25-671 Supplier Tocris Bioscience™ Supplier No. 2567/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Encephalitogenic myelin proteolipid fragment
PLP (139-151) is a synthetic myelin proteolipid protein (PLP) fragment. Immunization with this peptide induces severe clinical and histological experimental allergic encephalomyelitis (EAE), an animal model of multiple sclerosis.
Specifications
Quantity 1 mg