Learn More
Tocris Bioscience™ ProTx II
GREENER_CHOICE
Catalog No. 4023100U
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
40-231-00U 100 μg
1 options
This item is not returnable. View return policy
Catalog No. 40-231-00U Supplier Tocris Bioscience™ Supplier No. 4023/100U
Add to Cart
Add to Cart
This item is not returnable. View return policy
Selective NaV1.7 channel blocker
ProTx II is a selective NaV1.7 channel blocker. Shifts activation gating positively and decreases current magnitude. Displays 100-fold selectivity over other sodium channel subtypes.