Learn More
Tocris Bioscience™ RFRP 3 (human)
Catalog No. 46831
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
46-831 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 46-831 Supplier Tocris Bioscience™ Supplier No. 4683/1
This item is not returnable. View return policy
NPFF1 agonist; GnIH homolog
RFRP 3 (human) is an agonist of the NPFF1 receptor (IC50 = 0.7 nM for inhibition of forskolin-induced cAMP production). Homolog of gonadotropin-inhibitory hormone (GnIH); inhibits activity of gonadotropin-releasing hormone (GnRH) neurons.
Specifications
Purity 0.95
Quantity 1 mg