Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RGDS peptide
GREENER_CHOICE

Catalog No. 349810
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-981-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 34-981-0 Supplier Tocris Bioscience™ Supplier No. 3498/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Integrin binding sequence; inhibits integrin receptor function

RGDS peptide is an integrin binding sequence that inhibits integrin receptor function. Decreases systemic inflammation via inhibition of collagen-triggered activation of leukocytes and attenuates expression of inflammatory cytokines, iNOS and MMP-9. Promotes cell attachment and abrogates apoptosis via the mitochondrial pathway in osteoblasts in vitro.

Specifications

Purity 95%
Content And Storage Store at -20°C
Quantity 10 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.