Learn More
Tocris Bioscience™ [Sar9,Met(O2)11]-Substance P
GREENER_CHOICE
Catalog No. 11781
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
11781 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 11781 Supplier Tocris Bioscience™ Supplier No. 1178/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Potent and selective NK1 agonist
[Sar9,Met(O2)11]-Substance P is a potent selective NK1 tachykinin receptor agonist.
Specifications
Molecular Weight (g/mol) M.W. (Observed): 1393.68
Quantity 1 mg