Learn More
Tocris Bioscience™ SHU 9119
GREENER_CHOICE
Catalog No. 34201
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
34-201 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 34-201 Supplier Tocris Bioscience™ Supplier No. 3420/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
MC3 and MC4 antagonist; MC5 partial agonist
SHU 9119 is a potent melanocortin MC3 and MC4 receptor antagonist (IC50 values are 0.23 and 0.06 nM respectively) and MC5 partial agonist (EC50 = 0.12 nM). Upregulates expression of genes promoting lipogenesis and triglyceride storage (SCD1, LPL, ACCα and FAS), increases triglyceride synthesis and promotes insulin resistance. Increases food intake, body weight and fat mass when administered centrally in vivo.
Specifications
Purity 0.95
Quantity 1 mg