Learn More
Tocris Bioscience™ SNX 482
Catalog No. 294510U
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
29-4510U 10 μg
1 options
This item is not returnable. View return policy
Catalog No. 29-4510U Supplier Tocris Bioscience™ Supplier No. 2945/10U
This item is not returnable. View return policy
Potent and selective CaV2.3 blocker
SNX 482 is a potent and selective, voltage-dependent R-type CaV2.3 calcium channel blocker (IC50 = 30 nM). Antinociceptive; inhibits nociceptive C-fibre and Aδ-fibre-evoked neuronal responses.
Specifications
Quantity 10 μg