Learn More
Tocris Bioscience™ 10Panx
GREENER_CHOICE
Catalog No. 33481
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
33-481 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 33-481 Supplier Tocris Bioscience™ Supplier No. 3348/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Panx-1 mimetic inhibitory peptide; blocks pannexin-1 gap junctions
10Panx is a panx-1 mimetic inhibitory peptide that blocks pannexin-1 gap junctions. Inhibits P2X7-mediated dye uptake, ATP-mediated IL-1β release and caspase-1 activation without altering membrane current in macrophages in vitro. Also blocks activation of NMDA receptor secondary currents (I2nd) by > 70%.
Specifications
Purity 95%
Quantity 1 mg