Learn More
Tocris Bioscience™ TC 14012
Catalog No. 43001
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
43-001 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 43-001 Supplier Tocris Bioscience™ Supplier No. 4300/1
This item is not returnable. View return policy
CXCR4 antagonist; ACKR3 (CXCR7) agonist
TC 14012 is a CXCR4 antagonist and ACKR3 (CXCR7) agonist (EC50 = 350 nM for CXCR7).
Specifications
Purity 0.95
Quantity 1 mg