Learn More
Tocris Bioscience™ Urotensin II (human)
Catalog No. 16421
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
16-421 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 16-421 Supplier Tocris Bioscience™ Supplier No. 1642/1
This item is not returnable. View return policy
Endogenous vasoactive agonist for the UT receptor
Urotensin II (human) is a potent endogenous peptide agonist for the urotensin-II receptor (EC50 = 0.1 nM). Displays arterio-selective vasoconstriction and vasodilatation in mammals in vitro and in vivo, effects which vary between species. Also has been shown to mediate bronchoconstriction.
Specifications
Quantity 1 mg