Learn More
Tocris Bioscience™ VIP (human, rat, mouse, rabbit, canine, porcine)
GREENER_CHOICE
Catalog No. 19111
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
19-111 1 mg
1 options
This item is not returnable. View return policy
Catalog No. 19-111 Supplier Tocris Bioscience™ Supplier No. 1911/1
Add to Cart
Add to Cart
This item is not returnable. View return policy
Involved in neurotransmission and smooth muscle relaxation
VIP (human, rat, mouse, rabbit, canine, porcine) is a neuropeptide with many biological actions; plays a role in neurotransmission, smooth muscle relaxation and has trophic and mitogenic actions.
Specifications
Quantity 1 mg