Learn More
Thermo Scientific™ Todd Hewitt Broth
Description
Cultivate beta hemolytic streptococci, encourage the production of antigenic streptococci hemolysins, and isolate pathogenic microorganisms using convenient, pre-filled, ready-to-use Thermo Scientific™ Todd Hewitt Broth.
Streptococci are facultatively anaerobic, catalase-negative, gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fascitis. One of the most useful phenotypic characteristics of streptococci is the hemolytic reaction, generally classified as alpha, beta or gamma according to the appearance of zones around colonies growing on Blood Agar. Beta-hemolytic streptococci are further characterized by Lancefield groups based on the antigenic differences in group-specific polysaccharides located in the bacterial cell wall.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Clear liquid, light amber |
| Content And Storage | 2°C to 8°C |
| Description | Todd Hewitt Broth |
| Format | Tube |
| Manufacturing Origin | United States |
| Product Type | Microbiology Media |
| Fill Volume | 5 mL |
| Packaging Type | Glass tube with black screw cap |
| Quantity | 100/Pk. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.