Learn More
Grainger Toggle Valve 4 Way 1/8 In NPT
Supplier:  Grainger 14F282
Encompass_Preferred
The Fisherbrand 4-Way Toggle Valve, 1/8" NPT is a 4-way toggle valve with 1/8" NPT female ports. It is made of chrome-plated brass with a toggle handle. This valve allows flow between four ports and is suitable for directing flow between multiple lines. The valve has a maximum pressure rating of 150 psi and temperature range of -40 to 200°F. The valve body measures approximately 1-3/4" x 1-1/2" x 1-3/8". This 4-way toggle valve can be used for a variety of liquid and gas flow control applications.
Catalog No. 50-275-1002
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.