Learn More
Sigma Aldrich Fine Chemicals Biosciences TokeOni100MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 808350
Encompass
Luciferin is a generic term for the light-emitting compound found in organisms that generate bioluminescence.TokeOni is a new NIR emission luciferin analogue having luminescence peak at 670 ~ 680 nm with a peak range in the near-infrared (NIR) region.TokeOni has the enhanced solubility of 10 mg/mL compared to HCl free (0.2 mg/mL).
Catalog No. 50-290-6835