Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, ACS Reagent, for HPLC, Spectrophotometry, meets analytical spec. of USP, >99.8%, Solstice

Catalog No. 6001720 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
Poly-coated Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-017-20 4 x 4 L Glass Bottle
60-017-21 4 x 4 L Poly-coated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-017-20 Supplier Solstice Supplier No. AH3474
Add to Cart
Add to Cart

Chemical Identifiers

CAS 108-88-3
Molecular Formula C7H8
Molecular Weight (g/mol) 92.14
MDL Number MFCD00008512
InChI Key YXFVVABEGXRONW-UHFFFAOYSA-N
Synonym methylbenzene, toluol, phenylmethane, benzene, methyl, methylbenzol, methacide, toluen, antisal 1a, tolu-sol, monomethyl benzene
PubChem CID 1140
ChEBI CHEBI:17578
IUPAC Name toluene
SMILES CC1=CC=CC=C1

Specifications

Absorbance 0.1 AU max. at 300 nm, 0.025 AU max. at 325 nm
CAS Max % 100.0000%
Density 0.867 g/cm3
Quantity 4 x 4 L
Flash Point 4°C
Packaging Glass Bottle
UN Number UN1294
Water 0.03% max.
Beilstein 635760
Boiling Point 110.6°C
Solubility Information 5.2% in Water at 20°C
Formula Weight 92.14
Percent Purity >99.8%
Grade HPLC
Chemical Name or Material Toluene, ACS Reagent, for HPLC, Spectrophotometry, meets analytical specification of USP, >99.8%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.