Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Toluene, ACS Reagent, Reagents

Catalog No. 01337644
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
Packaging:
Glass Bottle
Safety Coated Glass
Steel Pail
Tin Can
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-337-644 4 L Safety Coated Glass
01-337-646 20 L Steel Pail
01-337-645 4 L Glass Bottle
01-337-643 4 L Tin Can
01-337-642 500 mL Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 01-337-644 Supplier Reagents Holdings Llc Supplier No. C13410004C1
Add to Cart
Add to Cart

Toluene, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Toluene, ACS Reagent
Melting Point -95°C
CAS 108-88-3
Color Colorless
Quantity 4 L
Packaging Safety Coated Glass
Physical Form Liquid
Specific Gravity 0.866
Molecular Formula C6H5CH3
Synonym Methylbenzene, Phenylmethane, Toluol
Boiling Point 111°C
Molecular Weight (g/mol) 92.1384
Grade ACS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.