Learn More
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -93°C (literature) |
| Absorbance | max. 1.00 at 286 nm, max. 0.01 at 400 nm, max. 0.01 at 350 nm, max. 0.10 at 300 nm, max. 0.50 at 288 nm |
| CAS Max % | 100.0000% |
| Density | 0.865 g/mL at 25 °C(lit.) |
| Quantity | 100 mL |
| Flash Point | 4°C |
| Packaging | Glass Bottle |
| Linear Formula | C6H5CH3 |
| UN Number | UN1294 |
| Beilstein | 635760 |
| Show More |
Safety and Handling
| ShelfLife | 1800 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Toluene |
| EINECSNumber | 203-625-9 |
| Recommended Storage | Room Temp |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.