Learn More
Sigma Aldrich Fine Chemicals Biosciences Toluene suitable for HPLC
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 34866
Encompass
Toluene, a benzene analog, is widely employed as an organic solvent. It has been reported to be a biotoxic solvent (toxic to many microorganisms at 0.1%v/v concentrations). Its anaerobic biodegradation to CO2, by the denitrifying bacterium Thauera arornatica has been reported. It forms a syndiotactic polystyrene-toluene molecular compound. Crystal structure of this molecular compound has been investigated by X-ray diffraction studies.
Catalog No. 50-288-6459