Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tolylene-2,6-diisocyanate 98.0+%, TCI America™
SDP

Catalog No. T115325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T115325G 25 g
T11531G 1 g
T11535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. T115325G Supplier TCI America Supplier No. T115325G
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 91-08-7
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.159
MDL Number MFCD00002010
InChI Key RUELTTOHQODFPA-UHFFFAOYSA-N
Synonym 2,6-diisocyanatotoluene, toluene-2,6-diisocyanate, 2,6-toluene diisocyanate, 2-methyl-m-phenylene diisocyanate, 2,6-tdi, m-tolylene diisocyanate, meta-tolylene diisocyanate, toluene 2,6-diisocyanate, benzene, 1,3-diisocyanato-2-methyl, 2,6-diisocyanato-1-methylbenzene
PubChem CID 7040
ChEBI CHEBI:53557
IUPAC Name 1,3-diisocyanato-2-methylbenzene
SMILES CC1=C(C=CC=C1N=C=O)N=C=O

Specifications

Color Yellow
Boiling Point 103°C
Quantity 25 g
UN Number 2078
Formula Weight 174.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Tolylene-2,6-diisocyanate
EINECSNumber (3)-2214
RTECSNumber CZ6310000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.