Learn More
TOM1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301362A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene was identified as a target of the v-myb oncogene. The encoded protein shares its N-terminal domain in common with proteins associated with vesicular trafficking at the endosome. It is recruited to the endosomes by its interaction with endofin. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.Specifications
| TOM1 | |
| Polyclonal | |
| Unconjugated | |
| TOM1 | |
| Target of myb1 (chicken), FLJ33404 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10043 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O60784 | |
| TOM1 | |
| Between 450 and 492 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.