Learn More
Invitrogen™ TOMM22 Recombinant Rabbit Monoclonal Antibody (7C11)

Description
This antibody has been tested in direct-ELISA.
Specifications
Specifications
| Antigen | TOMM22 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 7C11 |
| Concentration | 0.41 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol, 150mM NaCl and 0.02% sodium azide; pH 7.4 |
| Gene | Tomm22 |
| Gene Accession No. | Q9NS69 |
| Gene Alias | 1C9-2; 2310047D01; hTom22; Mitochondrial import receptor subunit TOM22 homolog; mitochondrial import receptor Tom22; MST065; MSTP065; rTOM22; Tom22; Tomm22; Translocase of outer membrane 22 kDa subunit homolog; translocase of outer mitochondrial membrane 22; translocase of outer mitochondrial membrane 22 homolog; translocase of outer mitochondrial membrane 22 homolog (yeast) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.