Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TOMM40 Mouse anti-Human, Mouse, Porcine, Rat, Clone: 5E2C5, Proteintech
SDP

Catalog No. 501737076 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7076 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7076 Supplier Proteintech Group Inc Supplier No. 666581IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.

Specifications

Antigen TOMM40
Applications Western Blot
Classification Monoclonal
Clone 5E2C5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene TOMM40
Gene Accession No. O96008, Q75Q40, Q9QYA2
Gene Alias C19orf1, D19S1177E, p38.5, PER EC1, PEREC1, Protein Haymaker, TOM40, TOMM40
Gene Symbols Tomm40
Host Species Mouse
Immunogen TOMM40 Fusion Protein Ag13065
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 102165927, 10452, 308416, 53333
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.