Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Tomosyn Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565579 Supplier Invitrogen™ Supplier No. PA565579
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SAHVIIYRFSK QEVITEVIPM LEVRLLYEIN DVETPEGEQP PPLPTPVGGS NPQPIPPQSH PSTSSSSSDG LRDN Highest antigen sequence identity to the following orthologs - mouse 95%, rat 93%.

Syntaxin 1 is a component of the 7S and 20S SNARE complexes which are involved in docking and fusion of synaptic vesicles with the presynaptic plasma membrane. This gene encodes a syntaxin 1 binding protein. In rat, a similar protein dissociates syntaxin 1 from the Munc18/n-Sec1/rbSec1 complex to form a 10S complex, an intermediate which can be converted to the 7S SNARE complex. Thus this protein is thought to be involved in neurotransmitter release by stimulating SNARE complex formation. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Tomosyn
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STXBP5
Gene Accession No. Q5T5C0
Gene Alias 0710001E20Rik; 4930565N16Rik; AW742610; FLJ30922; Kiaa4253; L Nbla04300; Lethal(2) giant larvae protein homolog 3; LGL3; LLGL3; MGC141942; MGC141968; mKIAA1006; Nbla04300; putative protein product of Nbla04300; RP11-361F15.4; STXB5; Stxbp5; syntaxin binding protein 5; syntaxin binding protein 5 (tomosyn); syntaxin-binding protein 5; tomosyn; tomosyn 1; tomosyn-1
Gene Symbols STXBP5
Host Species Rabbit
Immunogen Recombinant Human Tomosyn. Recombinant protein control fragment (Product #RP-106244).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134957
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.