Learn More
United Scientific™ Tongs, Stainless Steel with PTFE Tips
SDPSCI_ED
Catalog No. S01786 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Sterility:
Non-sterile
Catalog No. Sterility
S01786 Non-sterile
1 options
Catalog No. S01786 Supplier United Scientific™ Supplier No. UNSSTON09NS
Add to Cart
Add to Cart
Safely move items using these tongs with a firm grip.
Stainless steel tongs measure 9 inches long, 3 inches of the tips are coated with PTFE.
  • Available in sterile or non-sterile versions.
  • Autoclavable
  • 304 Stainless Steel
  • PTFE-coated tips
Specifications
Product Type Laboratory Tongs
Sterility Non-sterile
Material Stainless Steel
Length (English) 22.9 cm
Length (Metric) 9 in.