Learn More
Vikan™ Top Dog Sleeves, 18 in.
Catalog No. 17227957 Shop All Remco Products
Change view
Click to view available options
Color:
Blue
Green
Red
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-957 Green
17-227-959 Red
17-227-961 Yellow
17-227-958 Blue
17-227-960 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-227-957 Supplier Vikan™ Supplier No. 84012
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sleeves are elastic at both ends to keeps them in place while providing coverage from the wrist to above the elbow.
  • Reusable, 6 mil thick
  • Color-coded
  • FDA-compliant
  • Protect the user from the wrist to above the elbow
  • Elastic at both ends of the sleeves keeps the sleeve securely in place throughout a shift
  • The sleeves work well with Top Dog aprons
  • All sleeves are packaged in pairs
Specifications
Product Type General Purpose Sleeve
Size Universal
Material Ether-Based Polyurethane
Color Green
Sterility Non-sterile
Disposable Reusable
Seam Type Welded
Cuff Style Elastic
Certifications/Compliance FDA Compliant