Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TOP1MT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104428 Supplier Invitrogen™ Supplier No. PA5104428
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TOP1MT.

This gene encodes a mitochondrial DNA topoisomerase that plays a role in the modification of DNA topology. The encoded protein is a type IB topoisomerase and catalyzes the transient breaking and rejoining of DNA to relieve tension and DNA supercoiling generated in the mitochondrial genome during replication and transcription. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TOP1MT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TOP1MT
Gene Accession No. Q6IM78, Q8R4U6, Q969P6
Gene Alias 2900052H09Rik; DNA topoisomerase 1, mitochondrial; DNA topoisomerase I; DNA topoisomerase I, mitochondrial; mitochondrial topoisomerase I; mitochondrial topoisomerase IB; TOP1M; TOP1MT; topoisomerase (DNA) I, mitochondrial; Topoisomerase I mitochondrial; Type IB Topoisomerase
Gene Symbols TOP1MT
Host Species Rabbit
Immunogen A synthesized peptide derived from human TOP1MT(Accession Q969P6), corresponding to amino acid residues H203-T253.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116447, 300029, 72960
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.