Learn More
Selleck Chemical LLC Alprostadil (PGE1) S1508-100mg
SDP
Supplier:  Selleck Chemical LLC S1508100MG
Encompass_Preferred
Alprostadil (PGE1) is used as a drug in the treatment of erectile dysfunction and has vasodilatory properties It is a Prostaglandin Analog and Prostaglandin E1 Agonist
Catalog No. 50-311-0234
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.