Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4,4'-sulfonylbis(2-methylphenol) | 16346-97-7 | MFCD00130281 | 278.32 g/mol | C14H14O4S | 10 MG
SDP

Supplier:  Medchemexpress LLC HYW17801110MG

Encompass_Preferred

Topoisomerase IIα-IN-9 (4,4'-sulfonylbis(2-methylphenol)) is a bisphenol compound that moderately inhibits human DNA topoisomerase II and reduces cell proliferation in selected cell lines. It is supplied for laboratory research; lot-specific purity, solubility, and storage conditions are provided in the product data sheet and Certificate of Analysis.

  • Moderate topoisomerase II inhibition (IC50: 571 μM).
  • Inhibits K562 cell growth (IC50: 117 μM) and CHO cell growth (IC50: 218 μM).
  • Molecular weight 278.32 g/mol and formula C14H14O4S.
  • CAS number 16346-97-7 for unambiguous identification.
  • Intended for research use; consult Certificate of Analysis for purity and storage.

Catalog No. 50-002-83250


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.