Learn More
Medchemexpress LLC 4,4'-sulfonylbis(2-methylphenol) | 16346-97-7 | MFCD00130281 | 97.0% | 278.32 g·mol-1 | C14H14O4S | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW17801125MG
Encompass_Preferred
Topoisomerase IIα-IN-9 is a bisphenol-derived small-molecule inhibitor of human DNA topoisomerase IIα intended for research use. Reported biochemical activity includes Topo II inhibition (IC50 ≈ 571 μM) and cell growth inhibitory activity in K562 and CHO cells (IC50 ≈ 117 μM and 218 μM, respectively). Molecular formula C14H14O4S and molecular weight 278.32 g·mol-1 are provided; consult the Certificate of Analysis for purity and storage conditions.
  • Moderate inhibitor of DNA topoisomerase IIα.
  • Exhibits cell growth inhibition in selected cell lines.
  • Provided as a characterized small molecule with formula and molecular weight.
  • Suitable for biochemical assays and cell-based research applications.
  • Certificate of Analysis available for purity and handling details.
Catalog No. 50-002-83152
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.