Learn More
Enzo Life Sciences Topotecan . hydrochloride (1mg). CAS: 119413-54-6
SDP
Supplier:  Enzo Life Sciences ALX350133M001
Encompass_Preferred
Potent antitumor agent. Inhibitor of DNA-topoisomerase. Analog of (S)-(+)-camptothecin (ALX-350-015). Used in chemotherapy of several different cancers. Alternative name: 9-[(Dimethylamino)methyl]-10-hydroxy-(20S)-camptothecin . HCl. Purity: ≥98% (HPLC). Solubility: Soluble in water (1mg/ml) or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8750
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.