Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Torc1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582786
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582786 Supplier Invitrogen™ Supplier No. PA582786
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SPPADTSWRR TNSDSALHQS TMTPTQPESF SSGSQDVHQK RVLLLTVPGM EETTSEADKN LSKQAWDTKK TGSRPKSCEV Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 86%.

Transcriptional coactivator for CREB1 which activates transcription through both consensus and variant cAMP response element (CRE) sites. Acts as a coactivator, in the SIK/TORC signaling pathway, being active when dephosphorylated and acts independently of CREB1 'Ser-133' phosphorylation. Enhances the interaction of CREB1 with TAF4. Regulates the expression of specific CREB-activated genes such as the steroidogenic gene, StAR. Potent coactivator of PGC1alpha and inducer of mitochondrial biogenesis in muscle cells. In the hippocampus, involved in late-phase long-term potentiation (L-LTP) maintenance at the Schaffer collateral-CA1 synapses. May be required for dendritic growth of developing cortical neurons. In concert with SIK1, regulates the light-induced entrainment of the circadian clock. In response to light stimulus, coactivates the CREB-mediated transcription of PER1 which plays an important role in the photic entrainment of the circadian clock. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Torc1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CRTC1
Gene Accession No. Q157S1, Q68ED7, Q6UUV9
Gene Alias AI413414; CH211-255D18.10; CH211-255D18.9; CREB regulated transcription coactivator 1; CREB regulated transcription coactivator 1b; CREB-regulated transcription coactivator 1; CREB-regulated transcription coactivator 1; CREB-regulated transcription coactivator 1b; CRTC1; crtc1b; FLJ14027; KIAA0616; MECT 1; Mect1; mKIAA0616; mucoepidermoid carcinoma translocated 1; mucoepidermoid carcinoma translocated protein 1; Mucoepidermoid carcinoma translocated protein 1 homolog; R74955; si:ch211-255d18.8; TORC1; TORC-1; transducer of CREB protein 1; transducer of regulated cAMP response element-binding protein (CREB) 1; transducer of regulated cAMP response element-binding protein 1; WAMTP1; zfTorc1
Gene Symbols CRTC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CRTC1. Recombinant protein control fragment (Product #RP-93576).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23373, 382056, 684527
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.