Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TORC1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501558401
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-155-8401 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-155-8401 Supplier Bethyl Laboratories, Inc Supplier No. A300769AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with Tris-EDTA pH 9.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to

Transcriptional coactivator for CREB1 which activates transcription through both consensus and variant cAMP response element (CRE) sites.Acts as a coactivator, in the SIK/TORC signaling pathway,being active when dephosphorylated and acts independently of CREB1 ′Ser-133′ phosphorylation.Enhances the interaction of CREB1 with TAF4.Regulates the expression of specific CREB-activated genes such as the steroidogenic gene,StAR.Potent coactivator of PGC1alpha and inducer of mitochondrial biogenesis in muscle cells.Also coactivator for TAX activation of the human T-cell leukemia virus type 1 (HTLV-1) long terminal repeats (LTR).In the hippocampus, involved in late-phase long-term potentiation (L-LTP) maintece at the Schaffer collateral-CA1 synapses.May be required for dendritic growth of developing cortical neurons.

Specifications

Antigen TORC1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CRTC1
Gene Accession No. Q68ED7, Q6UUV9
Gene Alias CREB-regulated transcription coactivator 1; TORC-1; mucoepidermoid carcinoma translocated 1; mucoepidermoid carcinoma translocated protein 1; transducer of CREB protein 1; transducer of regulated cAMP response element-binding protein (CREB) 1; transducer of regulated cAMP response element-binding protein 1
Gene Symbols Crtc1
Host Species Rabbit
Immunogen Between 600 and C-term
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23373, 382056
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.