Learn More
Perkin Elmer US LLC SMARTintro Sample Introduction Module, Blue, with One-Piece 2.0 mm I.D. Quartz Torch-Injector
Supplier:  Perkin Elmer US LLC N8152437
Encompass_Preferred
This module, incorporated in the NexION 2000 Sample Introduction Base Configuration Kit (N8150034), includes: Cassette Piece (Blue) (N8152430) One-Piece Quartz Torch with 2 mm I.D. Injector (N8152472) Torch Injector Nut (N8152451) PTFE Torch Light Shield (N8152377) Torch Injector Removal Tool (N8152443)
Catalog No. 50-211-1619
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.