Learn More
Selleck Chemical LLC Torsemide 10mg 56211-40-6 AC-4464, JDL-464
SDP
Supplier:  Selleck Chemical LLC S169810MG
Encompass_Preferred
Torsemide (AC-4464, JDL-464) is a pyridyl sulfonylurea with a chemical structure between that of traditional loop diuretics and Cl- channel blockers, used to treat hypertension. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-656-0