Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Torsemide 98.0+%, TCI America™
SDP

Catalog No. T25381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T25381G 1 g
T25385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T25381G Supplier TCI America Supplier No. T25381G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56211-40-6
Molecular Formula C16H20N4O3S
Molecular Weight (g/mol) 348.421
MDL Number MFCD00866166
InChI Key NGBFQHCMQULJNZ-UHFFFAOYSA-N
Synonym N-(Isopropylcarbamoyl)-4-(m-tolylamino)pyridine-3-sulfonamide, Torasemide
PubChem CID 41781
ChEBI CHEBI:9637
IUPAC Name 1-[4-(3-methylanilino)pyridin-3-yl]sulfonyl-3-propan-2-ylurea
SMILES CC1=CC(=CC=C1)NC2=C(C=NC=C2)S(=O)(=O)NC(=O)NC(C)C

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 1 g
Formula Weight 348.42
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Torsemide
RTECSNumber UT7938000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.