Learn More
Pyramex™ Torser™ Safety Eyewear
Catalog No. 19155326 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Tint:
Clear
Gray
Light Gray
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint
19-155-326 Gray
19-155-325 Clear
19-155-327 Light Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-326 Supplier Pyramex™ Supplier No. GB10020TM
This item is not returnable. View return policy
H2MAX ensures maximum clarity in the most extreme temperature and humidity conditions
  • Dielectric
  • Flexible rubber gasket
  • Ratchet elastic headband with quick-fit retainer
  • Passes D3: Droplet and Splash Test and D4: Dust Test
  • H2MAX creates a high performing, superior anti-fog coating that is durable enough to withstand repeated cleaning without losing adhesion
  • H2MAX also adds a layer to the lens that is abrasion, chemical, and UV-resistant
  • Specifications
    Lens Coating Anti-fog
    Certifications/Compliance ANSI Z87.1,CSA Z94.3
    Certifications/Compliance (Europe) EN-166
    Lens Tint Gray
    Frame Color Black
    Weight (Metric) 46.8 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate, TPE