Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Products™ Torso Dust Cover
SCI_ED

Catalog No. S13152 Shop All Walter Products Products
Change view
Click to view available options
Length (English):
11 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
S13152 11 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S13152 Supplier Walter Products™ Supplier No. DCA02
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Heavy-duty dust cover that protects life-size human torso models.

  • Heavy-duty
  • Features a transparent window
  • For life-size human torso models

Specifications

Color Blue
For Use With (Application) Covers and protects life-size human torso models
Height (English) 2 in.
Length (English) 11 in.
Weight (English) 1 lb.
Width (English) 7 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.