Learn More
Scientific Industries TORBAL FSB2 Chuck Clamp Torque Gauge, 17lbf.in x 0.01lbf.in (2Nm x 0.001Nm) with Built-in Wireless, MicroSD, USB, RS232, and I/O
Supplier:  Scientific Industries FSB2
Encompass_Preferred
17lbf.in x 0.01lbf.in (2Nm x 0.001Nm) The FSB samples at a rate of 1000Hz and guarantees optimum precision and accuracy at all times. Measurements are automatically saved to the removable microSD memory card or can be easily sent via USB to a laptop or a PC. Powerful nickel–metal hydride batteries keep the gauge charged up to 16 hours. Standard, Peak, Multi-Peak, and Fill-Meter functions guarantee optimum functionality and flexibility when taking measurements. Every function includes statistical analysis, onscreen graphs, and a histogram. Analog I/O ports allows for threshold signaling and control of external peripherals.
Catalog No. NC2480385
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.