Learn More
Cygnus Technologies Tosoh R40 And R28 Protein A Mix-N-Go ELISA Kit, Streamlined Sample Prep, Two-Site Immunoenzymetric Assay, 96 Well Plate
Supplier:  Cygnus Technologies F910
Encompass
The Tosoh R40 and R28 Protein A Mix-N-Go ELISA Kit is a sensitive immunoassay designed for quantifying Protein A ligands from specific chromatographic resins, crucial in monoclonal antibody purification. It features Mix-N-Go™ technology, simplifying sample prep without boiling or centrifugation, thus speeding up results. Ideal for research and manufacturing, this kit enhances efficiency while ensuring accuracy during quality control and product release. Designed by Cygnus Technologies, it stands out for its rapid and user-friendly approach.
Catalog No. NC1958188
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.