Learn More
Selleck Chemical LLC Kobe2602
SDP
Supplier:  Selleck Chemical LLC S65272MG
Encompass_Preferred
Kobe2602 is a selective Ras inhibitor that blocks H-Ras GTP binding to c-Raf-1 (Ki 149 M)
Catalog No. 50-000-02979
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.