Learn More
Abcam AMPKa Total and Phospho T172 In-Cell ELISA Kit (IR), 1 x 96 Tests
Supplier:  Abcam AB1512801X96TESTS
Encompass_Preferred
AMPKa Total and Phospho T172 In-Cell ELISA Kit (IR) is Cell-based (quantitative) ELISA for the measurement of AMPKa Total and Phospho T172 (IR) production in Mouse, Rat, Human in Suspension cells, Adherent cells samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2333418
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.